Please login to your online account to display your discounted pricing

5-Chloromethyl-2-oxazolidinone 98%, ACROS Organics™

$267.98 - $267.98

Chemical Identifiers

Chemical Name or Material 5-Chloromethyl-2-oxazolidinone
Name Note 98%
CAS 22625-57-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H6ClNO2
Formula Weight 135.55
MDL Number MFCD00005269
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC159290050

  SDS
thermo fisher scientific
159290050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC159290250

  SDS
thermo fisher scientific
159290250
Glass bottle 25g Each for $267.98
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 135.55
Physical Form Crystalline Powder
Quantity 5g
Color White
Beilstein 27, 145
EINECS Number 245-137-9
Melting Point 101° to 105°C
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 5-Chloromethyl-2-oxazolidinone
Name Note 98%
Assay Percent Range 98%
Molecular Formula C4H6ClNO2
MDL Number MFCD00005269
CAS 22625-57-6
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.