Please login to your online account to display your discounted pricing

5-(Chloromethyl)-1,3-dimethyl-1h-pyrazole, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC12524DE

 View more versions of this product

Catalog No. CC12524DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 144.6
Quantity 5g
Assay 97%
Assay Percent Range 97%
CAS 852227-86-2
Chemical Name or Material 5-(Chloromethyl)-1, 3-dimethyl-1H-pyrazole
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Molecular Formula C6H9ClN2
Packaging Amber Glass Bottle
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. Harmful if swallowed.

Specifications

Chemical Name or Material 5-(Chloromethyl)-1, 3-dimethyl-1H-pyrazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H9ClN2
Formula Weight 144.6
Accessories

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

GHS P Statement Causes severe skin burns and eye damage. Harmful if swallowed.