Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloroindole, 98%

$76.54 - $917.62

Chemical Identifiers

Chemical Name or Material 5-Chloroindole
CAS 17422-32-1
Molecular Formula C8H6ClN
Formula Weight 151.6
MDL Number MFCD00005672
Beilstein 2651
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1359203

  SDS
alfa aesar™
A1359203
1g Each for $76.54

AAA1359206

  SDS
alfa aesar™
A1359206
5g Each for $235.05

AAA1359214

  SDS
alfa aesar™
A1359214
25g Each for $917.62
Description & Specifications

Specifications

Formula Weight 151.6
Chemical Name or Material 5-Chloroindole
Melting Point 69° to 73°C
Beilstein 2651
Health Hazard 1 Warning
Molecular Formula C8H6ClN
MDL Number MFCD00005672
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 17422-32-1
Assay 98%

Applications
5-Chloroindole has been used in the synthesis of 5-chloro-3-indole-N,N- dimethylglyoxalamide and 5-chloro-N,N-dimethyltryptamine. It may be used in the synthesis of dyestuffs in the presence of biocatalysts. 5-Chloroindole can be synthesized by using 3-chlorobenzaldehyde as starting reagent

Solubility
Soluble in alcohol.

Notes
Store at room temperature. Incompatible with acids, reducing agents and amines.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.