Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloroindole-3-carboxaldehyde, 98%

$56.50 - $173.28

Chemical Identifiers

Chemical Name or Material 5-Chloroindole-3-carboxaldehyde
CAS 827-01-0
Molecular Formula C9H6ClNO
Formula Weight 179.61
MDL Number MFCD00175291
Beilstein 123794
Synonym 5-Chloro-3-formylindole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2486503

  SDS
alfa aesar™
B2486503
1g Each for $56.50

AAB2486506

  SDS
alfa aesar™
B2486506
5g Each for $173.28
Description & Specifications

Specifications

Formula Weight 179.61
CAS 827-01-0
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
MDL Number MFCD00175291
Beilstein 123794
Synonym 5-Chloro-3-formylindole
Melting Point 210° to 212°C
Chemical Name or Material 5-Chloroindole-3-carboxaldehyde
Molecular Formula C9H6ClNO
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.