missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloroindole-2-carboxylic Acid 98.0+%, TCI America™
$607.68 - $607.68
Chemical Identifiers
| CAS | 10517-21-2 |
|---|---|
| Molecular Formula | C9H5ClNO2 |
| Molecular Weight (g/mol) | 194.59 |
| MDL Number | MFCD00005613 |
| InChI Key | FUQOTYRCMBZFOL-UHFFFAOYSA-M |
| Synonym | 5-chloroindole-2-carboxylic acid, 5-chloroindole-2-carboxylate, 1h-indole-2-carboxylic acid, 5-chloro, spectrum_000183, 5-chloroindoline-2-carboxylicacid, pubchem1670, acmc-1bztj, specplus_000670, spectrum2_000513, spectrum3_000783 |
| PubChem CID | 82693 |
| IUPAC Name | 5-chloro-1H-indole-2-carboxylate |
| SMILES | [O-]C(=O)C1=CC2=CC(Cl)=CC=C2N1 |
Chemical Identifiers
| 10517-21-2 | |
| 194.59 | |
| FUQOTYRCMBZFOL-UHFFFAOYSA-M | |
| 82693 | |
| [O-]C(=O)C1=CC2=CC(Cl)=CC=C2N1 |
| C9H5ClNO2 | |
| MFCD00005613 | |
| 5-chloroindole-2-carboxylic acid, 5-chloroindole-2-carboxylate, 1h-indole-2-carboxylic acid, 5-chloro, spectrum_000183, 5-chloroindoline-2-carboxylicacid, pubchem1670, acmc-1bztj, specplus_000670, spectrum2_000513, spectrum3_000783 | |
| 5-chloro-1H-indole-2-carboxylate |
Specifications
| 10517-21-2 | |
| White-Yellow | |
| MFCD00005613 | |
| 5-chloroindole-2-carboxylic acid, 5-chloroindole-2-carboxylate, 1h-indole-2-carboxylic acid, 5-chloro, spectrum_000183, 5-chloroindoline-2-carboxylicacid, pubchem1670, acmc-1bztj, specplus_000670, spectrum2_000513, spectrum3_000783 | |
| [O-]C(=O)C1=CC2=CC(Cl)=CC=C2N1 | |
| 194.59 | |
| 195.60 | |
| Crystalline Powder |
| 290°C | |
| C9H5ClNO2 | |
| 25 g | |
| FUQOTYRCMBZFOL-UHFFFAOYSA-M | |
| 5-chloro-1H-indole-2-carboxylate | |
| 82693 | |
| ≥98.0% (GC,T) | |
| 5-Chloroindole-2-carboxylic Acid |
Safety and Handling
RTECSNumber : NL5998400
TSCA : No