Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloroindole-2-carboxylic acid, 98%

$204.92 - $204.92

Chemical Identifiers

Chemical Name or Material 5-Chloroindole-2-carboxylic acid
CAS 10517-21-2
Molecular Formula C9H6ClNO2
Formula Weight 195.6
MDL Number MFCD00005613
Beilstein 153229
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1862606

  SDS
alfa aesar™
A1862606
5g Each for $204.92
Description & Specifications

Specifications

Formula Weight 195.6
Chemical Name or Material 5-Chloroindole-2-carboxylic acid
MDL Number MFCD00005613
Molecular Formula C9H6ClNO2
Assay 98%
Beilstein 153229
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Melting Point 287°C (dec.)
RTECS Number NL5998400
CAS 10517-21-2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.