missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chlorobenzoxazole, 95%

$83.32 - $83.32

Chemical Identifiers

CAS 17200-29-2
Molecular Formula C7H4ClNO
Formula Weight 153.57
MDL Number MFCD00216939
Chemical Name or Material 5-Chlorobenzoxazole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2708306 SDS Alfa Aesar™
H2708306
5g Each for $83.32
Specifications Description & Specifications

Chemical Identifiers

17200-29-2
153.57
5-Chlorobenzoxazole
C7H4ClNO
MFCD00216939

Specifications

39° to 41°C
17200-29-2
153.57
5-Chlorobenzoxazole
5g
C7H4ClNO
MFCD00216939
95%

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning