missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chlorobenzofuroxan, 97%

$80.78 - $311.15

Chemical Identifiers

CAS 17348-69-5
Molecular Formula C6H3ClN2O2
Formula Weight 170.56
MDL Number MFCD00068063
Synonym 5-Chlorobenzofurazan-1-oxide
Chemical Name or Material 5-Chlorobenzofuroxan
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1426106 Alfa Aesar™
A1426106
5g Each for $80.78
AAA1426114 Alfa Aesar™
A1426114
25g Each for $311.15
Specifications Description & Specifications

Chemical Identifiers

17348-69-5
170.56
5-Chlorobenzofurazan-1-oxide
C6H3ClN2O2
MFCD00068063
5-Chlorobenzofuroxan

Specifications

45° to 48°C
17348-69-5
170.56
5-Chlorobenzofurazan-1-oxide
97%
5g
C6H3ClN2O2
MFCD00068063
5-Chlorobenzofuroxan