Please login to your online account to display your discounted pricing

5-Chloro-o-anisic acid 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Chloro-o-anisic acid
CAS 3438-16-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H7ClO3
Linear Formula CH3OC6H3(Cl)CO2H
Formula Weight 186.59
MDL Number MFCD00017546
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC393460050

  SDS
thermo fisher scientific
AC393460050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC393460250

  SDS
thermo fisher scientific
AC393460250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 186.59
Quantity 5g
Color White to off-white
Melting Point 96° to 99°C
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 5-Chloro-o-anisic acid
Linear Formula CH3OC6H3(Cl)CO2H
CAS 3438-16-2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS Min % 98.5
Molecular Formula C8H7ClO3
MDL Number MFCD00017546
Infrared Spectrum Authentic
Assay Percent Range 99%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.