Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-8-hydroxy-7-iodoquinoline, 98%

$53.73 - $717.31

Chemical Identifiers

Chemical Name or Material 5-Chloro-8-hydroxy-7-iodoquinoline
CAS 130-26-7
Molecular Formula C9H5ClINO
Formula Weight 305.49
MDL Number MFCD00006787
Beilstein 153637
Merck Index 14,5031
Synonym Clioqinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1113709

  SDS
alfa aesar™
A1113709
10g Each for $53.73

AAA1113718

  SDS
alfa aesar™
A1113718
50g Each for $183.18

AAA1113730

  SDS
alfa aesar™
A1113730
250g Each for $717.31
Description & Specifications

Specifications

Formula Weight 305.49
Chemical Name or Material 5-Chloro-8-hydroxy-7-iodoquinoline
TSCA TSCA
Assay 98%
Molecular Formula C9H5ClINO
MDL Number MFCD00006787
CAS 130-26-7
Beilstein 153637
Merck Index 14,5031
Synonym Clioqinol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H361-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P342+P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number VC5075000
Melting Point ≈175°C (dec.)
Quantity 10g

Applications
Clioquinol is a metal chelating ligand which displays protective effects in various neurodegenerative states as well as being a component in antifungal and antiprotozoal agents. Clioquinol rescues lead-induced impairments of synaptic plasticity in an animal models

Solubility
Soluble in DMSO (>25 mg/ml), boiling alcohol ((1:43)), methanol, and chloroform ((1:120)).

Notes
Keep container tightly closed. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P342+P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H361-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
May cause an allergic skin reaction.