missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-8-hydroxy-7-iodoquinoline, 98%

Manufacturer:  Alfa Aesar™ A1113718

 View more versions of this product

Catalog No. AAA1113718



Description

Description

Applications
Clioquinol is a metal chelating ligand which displays protective effects in various neurodegenerative states as well as being a component in antifungal and antiprotozoal agents. Clioquinol rescues lead-induced impairments of synaptic plasticity in an animal models

Solubility
Soluble in DMSO (>25 mg/ml), boiling alcohol ((1:43)), methanol, and chloroform ((1:120)).

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

5-Chloro-8-hydroxy-7-iodoquinoline
~175°C (decomposition)
130-26-7
MFCD00006787
14,5031
Soluble in DMSO (>25mg/ml),boiling alcohol ((1:43)),methanol,and chloroform ((1:120)).
C1=CC2=C(C(=C(C=C2Cl)I)O)N=C1
305.499
CHEBI:74460
98%
Light sensitive
50g
C9H5ClINO
153637
5-chloro-8-hydroxy-7-iodoquinoline, chinoform, chloroiodoquin, chloroiodoquine, chlorojodochin, clioquinol, cliquinol, iodochlorhydroxyquin, iodochloroxyquinoline, vioform
QCDFBFJGMNKBDO-UHFFFAOYSA-N
5-chloro-7-iodoquinolin-8-ol
2788
305.49

Chemical Identifiers

130-26-7
305.499
QCDFBFJGMNKBDO-UHFFFAOYSA-N
2788
5-chloro-7-iodoquinolin-8-ol
C9H5ClINO
MFCD00006787
5-chloro-8-hydroxy-7-iodoquinoline, chinoform, chloroiodoquin, chloroiodoquine, chlorojodochin, clioquinol, cliquinol, iodochlorhydroxyquin, iodochloroxyquinoline, vioform
CHEBI:74460
C1=CC2=C(C(=C(C=C2Cl)I)O)N=C1
Safety and Handling

Safety and Handling

GHS H Statement
H334-H361-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H361

EINECSNumber : 204-984-4

RTECSNumber : VC5075000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents