missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-6-nitroisoindoline-1,3-dione, ≥95%, Maybridge™

Chemical Identifiers

CAS 6015-57-2
Molecular Formula C8H3ClN2O4
Molecular Weight (g/mol) 226.572
MDL Number MFCD00052331
InChI Key ADLVDYMTBOSDFE-UHFFFAOYSA-N
Synonym 1h-isoindole-1,3 2h-dione, 5-chloro-6-nitro, 1h-isoindole-1,3 2h-dione,5-chloro-6-nitro, 4-chloro-5-nitrophthalimide, 5-chloro-6-nitro-2h-isoindole-1,3-dione, 5-chloro-6-nitro-isoindole-1,3-dione, 5-chloro-6-nitroindoline-1,3-dione, 5-chloro-6-nitroisoindoline-1,3-dione, 6-chloro-5-nitro-2h-benzo c azolidine-1,3-dione, 6-chloro-5-nitro-2h-benzo c azoline-1,3-dione, acmc-1b2sd
PubChem CID 2801129
IUPAC Name 5-chloro-6-nitroisoindole-1,3-dione
SMILES C1=C2C(=CC(=C1[N+](=O)[O-])Cl)C(=O)NC2=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
TL00400DA
SDS View Documents
Fisher Scientific Company
TL00400DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

6015-57-2
226.572
ADLVDYMTBOSDFE-UHFFFAOYSA-N
2801129
C1=C2C(=CC(=C1[N+](=O)[O-])Cl)C(=O)NC2=O
C8H3ClN2O4
MFCD00052331
1h-isoindole-1,3 2h-dione, 5-chloro-6-nitro, 1h-isoindole-1,3 2h-dione,5-chloro-6-nitro, 4-chloro-5-nitrophthalimide, 5-chloro-6-nitro-2h-isoindole-1,3-dione, 5-chloro-6-nitro-isoindole-1,3-dione, 5-chloro-6-nitroindoline-1,3-dione, 5-chloro-6-nitroisoindoline-1,3-dione, 6-chloro-5-nitro-2h-benzo c azolidine-1,3-dione, 6-chloro-5-nitro-2h-benzo c azoline-1,3-dione, acmc-1b2sd
5-chloro-6-nitroisoindole-1,3-dione

Specifications

Amber Glass Bottle
6015-57-2
95%
MFCD00052331
ADLVDYMTBOSDFE-UHFFFAOYSA-N
5-chloro-6-nitroisoindole-1,3-dione
2801129
95%
1g
95.0
C8H3ClN2O4
1h-isoindole-1,3 2h-dione, 5-chloro-6-nitro, 1h-isoindole-1,3 2h-dione,5-chloro-6-nitro, 4-chloro-5-nitrophthalimide, 5-chloro-6-nitro-2h-isoindole-1,3-dione, 5-chloro-6-nitro-isoindole-1,3-dione, 5-chloro-6-nitroindoline-1,3-dione, 5-chloro-6-nitroisoindoline-1,3-dione, 6-chloro-5-nitro-2h-benzo c azolidine-1,3-dione, 6-chloro-5-nitro-2h-benzo c azoline-1,3-dione, acmc-1b2sd
C1=C2C(=CC(=C1[N+](=O)[O-])Cl)C(=O)NC2=O
226.572
226.57
5-Chloro-6-nitroisoindoline-1, 3-dione
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS