missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H54432MD

 View more versions of this product

Catalog No. AAH54432MD



Description

Description

Applications
5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester
1083168-91-5
MFCD08063054
Slightly soluble in water.
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)OC)Cl
269.532
269.53
250mg
C12H17BClNO3
3-chloro-2-methoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine
ZHWIZWNYOSGXSR-UHFFFAOYSA-N
3-chloro-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
53398545
96%

Chemical Identifiers

1083168-91-5
269.532
ZHWIZWNYOSGXSR-UHFFFAOYSA-N
53398545
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)OC)Cl
C12H17BClNO3
MFCD08063054
3-chloro-2-methoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine
3-chloro-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents