Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-4-methyl-2-nitroaniline, 97%

$45.65 - $670.51

Chemical Identifiers

Chemical Name or Material 5-Chloro-4-methyl-2-nitroaniline
CAS 7149-80-6
Molecular Formula C7H7ClN2O2
Formula Weight 186.6
MDL Number MFCD00034066
Synonym 5-Chloro-2-nitro-p-toluidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1914803

  SDS
alfa aesar™
A1914803
1g Each for $45.65

AAA1914806

  SDS
alfa aesar™
A1914806
5g Each for $162.73

AAA1914814

  SDS
alfa aesar™
A1914814
25g Each for $670.51
Description & Specifications

Specifications

Formula Weight 186.6
Chemical Name or Material 5-Chloro-4-methyl-2-nitroaniline
Assay 97%
Molecular Formula C7H7ClN2O2
MDL Number MFCD00034066
CAS 7149-80-6
Synonym 5-Chloro-2-nitro-p-toluidine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 159° to 163°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.