Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-3-phenyl-2,1-benzisoxazole, 98+%

$34.32 - $294.00

Chemical Identifiers

Chemical Name or Material 5-Chloro-3-phenyl-2,1-benzisoxazole
CAS 719-64-2
Molecular Formula C13H8ClNO
Formula Weight 229.67
MDL Number MFCD00014573
Synonym 5-Chloro-3-phenylanthranil
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2313009

 
alfa aesar™
B2313009
10g Each for $34.32

AAB2313018

 
alfa aesar™
B2313018
50g Each for $78.04

AAB2313030

 
alfa aesar™
B2313030
250g Each for $294.00
Description & Specifications

Specifications

Formula Weight 229.67
Chemical Name or Material 5-Chloro-3-phenyl-2,1-benzisoxazole
TSCA TSCA
Assay ≥98%
Molecular Formula C13H8ClNO
MDL Number MFCD00014573
CAS 719-64-2
Synonym 5-Chloro-3-phenylanthranil
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 114° to 117°C
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.