missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-chloro-3-phenyl-2,1-benzisoxazole, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC438380250
Specifications
5-Chloro-3-phenyl-2, 1-benzisoxazole | |
Yellow | |
25g | |
96.0 | |
96% min. (GC) | |
2, 5-chloro-3-phenyl, 3-phenyl-5-chloro-2,1-benzisoxazole, 3-phenyl-5-chloroanthranil, 5-chloro-3-phenyl-2,1-benzisoxazole, 5-chloro-3-phenyl-benzo c isoxazole, 5-chloro-3-phenylanthranil, 5-chloro-3-phenylbenzo c isoxazole, acmc-209okx, maybridge1_004397, pubchem8713 | |
C1=CC=C(C=C1)C2=C3C=C(C=CC3=NO2)Cl | |
229.67 | |
229.67 | |
97% |
Authentic | |
115.3°C to 116.1°C | |
719-64-2 | |
100.0 | |
C13H8ClNO | |
MUHJZJKVEQASGY-UHFFFAOYSA-N | |
5-chloro-3-phenyl-2,1-benzoxazole | |
347291 | |
Crystalline Powder |
Chemical Identifiers
719-64-2 | |
229.67 | |
2, 5-chloro-3-phenyl, 3-phenyl-5-chloro-2,1-benzisoxazole, 3-phenyl-5-chloroanthranil, 5-chloro-3-phenyl-2,1-benzisoxazole, 5-chloro-3-phenyl-benzo c isoxazole, 5-chloro-3-phenylanthranil, 5-chloro-3-phenylbenzo c isoxazole, acmc-209okx, maybridge1_004397, pubchem8713 | |
5-chloro-3-phenyl-2,1-benzoxazole |
C13H8ClNO | |
MUHJZJKVEQASGY-UHFFFAOYSA-N | |
347291 | |
C1=CC=C(C=C1)C2=C3C=C(C=CC3=NO2)Cl |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
EINECSNumber : 211-950-2