Please login to your online account to display your discounted pricing

5-Chloro-3-methylbenzo[b]thiophene, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 5-Chloro-3-methylbenzo[b]thiophene
Assay Percent Range 97%
Molecular Formula C9H7ClS
Formula Weight 182.67
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC450990010

  SDS
thermo fisher scientific
AC450990010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC450990050

 
thermo fisher scientific
AC450990050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC450990250

  SDS
thermo fisher scientific
AC450990250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 182.67
Quantity 1g
Assay Percent Range 97%
Molecular Formula C9H7ClS
CAS 19404-18-3
Chemical Name or Material 5-Chloro-3-methylbenzo[b]thiophene