Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-3-methylbenzo[b]thiophene, 97%

$772.72 - $772.72

Chemical Identifiers

Chemical Name or Material 5-Chloro-3-methylbenzo[b]thiophene
CAS 19404-18-3
Molecular Formula C9H7ClS
Formula Weight 182.67
MDL Number MFCD00052506
Synonym 5-Chloro-3-methylthianaphthene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1182714

  SDS
alfa aesar™
A1182714
25g Each for $772.72
Description & Specifications

Specifications

Formula Weight 182.67
Chemical Name or Material 5-Chloro-3-methylbenzo[b]thiophene
Assay 97%
Molecular Formula C9H7ClS
MDL Number MFCD00052506
CAS 19404-18-3
Synonym 5-Chloro-3-methylthianaphthene
Melting Point 33° to 36°C
Quantity 25g