Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-3-methyl-1-(4-methylphenyl)-1H-pyrazole-4-carboxaldehyde, 96%

$101.28 - $347.88

Chemical Identifiers

Chemical Name or Material 5-Chloro-3-methyl-1-(4-methylphenyl)-1H-pyrazole-4-carboxaldehyde
CAS 350997-70-5
Molecular Formula C12H11ClN2O
Formula Weight 234.69
MDL Number MFCD01993680
Synonym 5-Chloro-3-methyl-1-(p-tolyl)-1H-pyrazole-4-carboxaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5048903

 
alfa aesar™
H5048903
1g Each for $347.88

AAH50489MD

 
alfa aesar™
H50489MD
250mg Each for $101.28
Description & Specifications

Specifications

Formula Weight 234.69
Chemical Name or Material 5-Chloro-3-methyl-1-(4-methylphenyl)-1H-pyrazole-4-carboxaldehyde
Molecular Formula C12H11ClN2O
MDL Number MFCD01993680
CAS 350997-70-5
Synonym 5-Chloro-3-methyl-1-(p-tolyl)-1H-pyrazole-4-carboxaldehyde
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Assay 96%
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.