missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H3306503

 View more versions of this product

Catalog No. AAH3306503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

5-Chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde
C9H8ClFO2
1g
VILWOSMVUDTFHT-UHFFFAOYSA-N
5-chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde
98001124
97%
1823049-90-6
MFCD22682805
Air sensitive
CC1=C(C(=C(C=C1C=O)Cl)OC)F
202.609
202.61

Chemical Identifiers

1823049-90-6
202.609
VILWOSMVUDTFHT-UHFFFAOYSA-N
5-chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde
C9H8ClFO2
MFCD22682805
98001124
CC1=C(C(=C(C=C1C=O)Cl)OC)F
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions