Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-(trifluoromethyl)phenylacetonitrile, 97%

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-(trifluoromethyl)phenylacetonitrile
CAS 261763-26-2
Molecular Formula C9H5ClF3N
Formula Weight 219.59
MDL Number MFCD01631357
UN Number UN3439
Synonym 5-Chloro-2-(trifluoromethyl)benzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2362903

  SDS
alfa aesar™
B2362903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2362906

  SDS
alfa aesar™
B2362906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 219.59
Health Hazard 2 GHS H Statement
H301-H312-H332-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Melting Point 61° to 63°C
Molecular Formula C9H5ClF3N
Chemical Name or Material 5-Chloro-2-(trifluoromethyl)phenylacetonitrile
MDL Number MFCD01631357
UN Number UN3439
Synonym 5-Chloro-2-(trifluoromethyl)benzyl cyanide
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
CAS 261763-26-2
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H312-H332-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.