Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-(trifluoromethyl)phenylacetic acid, 97%

$123.41 - $366.15

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-(trifluoromethyl)phenylacetic acid
CAS 261763-25-1
Molecular Formula C9H6ClF3O2
Formula Weight 238.59
MDL Number MFCD01631358
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2627203

  SDS
alfa aesar™
H2627203
1g Each for $123.41

AAH2627206

  SDS
alfa aesar™
H2627206
5g Each for $366.15
Description & Specifications

Specifications

Formula Weight 238.59
Molecular Formula C9H6ClF3O2
Assay 97%
Health Hazard 1 Warning
Chemical Name or Material 5-Chloro-2-(trifluoromethyl)phenylacetic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD01631358
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 117° to 119°C
CAS 261763-25-1
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.