Please login to your online account to display your discounted pricing

5-Chloro-2-thiophenecarboxaldehyde 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  405080050

 View more versions of this product

Catalog No. AC405080050

  • / Each
 

Description & Specifications

Chemical Identifiers

Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 2 GHS H Statement
May cause respiratory irritation. Harmful if swallowed. Harmful in contact with skin. Causes serious eye irritation. Harmful if inhaled. Causes skin irritation.
Assay ≥96.0% (GC)
Formula Weight 146.6
Physical Form Crystalline Powder or Solid
Quantity 5g
Color White to tan
EINECS Number 230-708-7
TSCA TSCA
CAS Min % 96.0
CAS Max % 100.0
Additional Information the substance becomes orange-red on aging
Health Hazard 1 Warning
MDL Number MFCD00047090
Chemical Name or Material 5-Chloro-2-thiophenecarboxaldehyde
CAS 7283-96-7
Assay Percent Range 97%
Name Note 97%
Molecular Formula C5H3ClOS
Packaging Glass bottle
Refractive Index 1.6040 to 1.6070 (20°C, 589nm)
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 5-Chloro-2-thiophenecarboxaldehyde
Name Note 97%
CAS 7283-96-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C5H3ClOS
Formula Weight 146.6
MDL Number MFCD00047090
Accessories

Warning

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
May cause respiratory irritation. Harmful if swallowed. Harmful in contact with skin. Causes serious eye irritation. Harmful if inhaled. Causes skin irritation.

SDS & Safety Documents