Learn More
5-Chloro-2-pentanone, 94%, Thermo Scientific Chemicals
$168.37 - $657.82
Chemical Identifiers
CAS | 5891-21-4 |
---|---|
Molecular Formula | C5H9ClO |
Molecular Weight (g/mol) | 120.576 |
MDL Number | MFCD00001008 |
InChI Key | XVRIEWDDMODMGA-UHFFFAOYSA-N |
Synonym | 5-chloro-2-pentanone, 2-pentanone, 5-chloro, 1-chloro-4-pentanone, 3-chloropropyl methyl ketone, 2-pentanone 5-chloro, 1-chloro-4-oxopentane, pubchem17207, 3-acetylpropyl chloride, 5-chloro-pentan-2-one, 5-chloranylpentan-2-one |
PubChem CID | 79993 |
IUPAC Name | 5-chloropentan-2-one |
SMILES | CC(=O)CCCCl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1026922
|
Thermo Scientific Chemicals
A1026922 |
100 g |
Each for $168.37
|
|
|||||
AAA1026936
|
Thermo Scientific Chemicals
A1026936 |
500 g |
Each for $657.82
|
|
|||||
Description
5-Chloro-2-pentanone used in the preparation of destructible surfactants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Chloro-2-pentanone used in the preparation of destructible surfactants.
Solubility
Soluble in chloroform and methanol.
Notes
keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents
Chemical Identifiers
5891-21-4 | |
120.576 | |
XVRIEWDDMODMGA-UHFFFAOYSA-N | |
79993 | |
CC(=O)CCCCl |
C5H9ClO | |
MFCD00001008 | |
5-chloro-2-pentanone, 2-pentanone, 5-chloro, 1-chloro-4-pentanone, 3-chloropropyl methyl ketone, 2-pentanone 5-chloro, 1-chloro-4-oxopentane, pubchem17207, 3-acetylpropyl chloride, 5-chloro-pentan-2-one, 5-chloranylpentan-2-one | |
5-chloropentan-2-one |
Specifications
5891-21-4 | |
63°C to 64°C (12 mmHg) | |
C5H9ClO | |
MFCD00001008 | |
UN1224 | |
5-chloro-2-pentanone, 2-pentanone, 5-chloro, 1-chloro-4-pentanone, 3-chloropropyl methyl ketone, 2-pentanone 5-chloro, 1-chloro-4-oxopentane, pubchem17207, 3-acetylpropyl chloride, 5-chloro-pentan-2-one, 5-chloranylpentan-2-one | |
XVRIEWDDMODMGA-UHFFFAOYSA-N | |
5-chloropentan-2-one | |
79993 | |
94% |
1.053 | |
35°C (95°F) | |
1.438 | |
100 g | |
1098490 | |
Soluble in chloroform and methanol. | |
CC(=O)CCCCl | |
120.576 | |
120.58 | |
5-Chloro-2-pentanone |
Safety and Handling
GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber : 227-565-8
RTECSNumber : SA8143500
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only