missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-pentanone, 94%

Manufacturer:  Alfa Aesar™ A1026936

 View more versions of this product

Catalog No. AAA1026936



Description

Description

Applications
5-Chloro-2-pentanone used in the preparation of destructible surfactants.

Solubility
Soluble in chloroform and methanol.

Notes
keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents
Specifications

Specifications

5-Chloro-2-pentanone
63°C to 64°C (12mmHg)
5891-21-4
C5H9ClO
UN1224
XVRIEWDDMODMGA-UHFFFAOYSA-N
5-chloropentan-2-one
79993
35°C (95°F)
500g
94%
MFCD00001008
1098490
CC(=O)CCCCl
120.576
120.58

Chemical Identifiers

5891-21-4
120.576
XVRIEWDDMODMGA-UHFFFAOYSA-N
5-chloropentan-2-one
C5H9ClO
MFCD00001008
79993
CC(=O)CCCCl
Safety and Handling

Safety and Handling

GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

RTECSNumber : SA8143500

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents