missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-2-nitrotoluene, 98%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 136640050

 View more versions of this product

Catalog No. AC136640050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

5-Chloro-2-nitrotoluene
5g
5367-28-2
100.0
Glass bottle
Liquid
98%
ClC6H3(NO2)CH3
MFCD00024579
Clear yellow
98%
97.5
Authentic
1.5700 to 1.5720 (20°C, 589nm)
≥97.5%
C7H6ClNO2
171.58
05, 327

Chemical Identifiers

5367-28-2
100.0
C7H6ClNO2
171.58
5-Chloro-2-nitrotoluene, 98%
97.5
98%
ClC6H3(NO2)CH3
MFCD00024579
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-355-3

RTECSNumber : XS9134000