Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-nitropyridine, 97%

$67.11 - $577.23

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-nitropyridine
CAS 52092-47-4
Molecular Formula C5H3ClN2O2
Formula Weight 158.54
MDL Number MFCD03092916
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2700003

 
alfa aesar™
H2700003
1g Each for $67.11

AAH2700006

 
alfa aesar™
H2700006
5g Each for $189.34

AAH2700014

 
alfa aesar™
H2700014
25g Each for $577.23
Description & Specifications

Specifications

Formula Weight 158.54
Chemical Name or Material 5-Chloro-2-nitropyridine
Assay 97%
Molecular Formula C5H3ClN2O2
MDL Number MFCD03092916
CAS 52092-47-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 120° to 123°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.