missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-nitrodiphenylamine, 98%

$38.76 - $394.92

Chemical Identifiers

CAS 25781-92-4
Molecular Formula C12H9ClN2O2
Formula Weight 248.67
MDL Number MFCD00007287
Chemical Name or Material 5-Chloro-2-nitrodiphenylamine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2417922 SDS Alfa Aesar™
B2417922
100g Each for $394.92
AAB2417906 SDS Alfa Aesar™
B2417906
5g Each for $38.76
AAB2417914 SDS Alfa Aesar™
B2417914
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

25781-92-4
248.67
5-Chloro-2-nitrodiphenylamine
C12H9ClN2O2
MFCD00007287

Specifications

108° to 111°C
25781-92-4
248.67
5-Chloro-2-nitrodiphenylamine
100g
C12H9ClN2O2
MFCD00007287
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning