Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-nitrobenzotrifluoride, 99%

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-nitrobenzotrifluoride
CAS 118-83-2
Molecular Formula C7H3ClF3NO2
Formula Weight 225.55
MDL Number MFCD00007298
UN Number UN2810
Beilstein 1973477
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1156106

  SDS
alfa aesar™
L1156106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1156114

  SDS
alfa aesar™
L1156114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.55
CAS 118-83-2
Molecular Formula C7H3ClF3NO2
Assay 99%
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
MDL Number MFCD00007298
UN Number UN2810
Quantity 5g
Boiling Point 222° to 224°C
Beilstein 1973477
Melting Point 21° to 22°C
Chemical Name or Material 5-Chloro-2-nitrobenzotrifluoride
Flash Point 102°C (215°F)
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.