Please login to your online account to display your discounted pricing

5-chloro-2-nitrobenzoic acid, 99%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC109685000

 View more versions of this product

Catalog No. AC109685000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 201.57
Physical Form Crystalline Powder
Quantity 500g
Color Light yellow to yellow-green
Melting Point 136° to 139°C
Beilstein 09, 401
CAS Max % 100.0
Loss on Drying 0.5% max.
Packaging Glass bottle
Molecular Formula C7H4ClNO4
Chemical Name or Material 5-Chloro-2-nitrobenzoic acid
Assay Percent Range 99%
CAS 2516-95-2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
MDL Number MFCD00007290
CAS Min % 98.5
Infrared Spectrum Authentic
Linear Formula ClC6H3(NO2)CO2H

Specifications

Chemical Name or Material 5-Chloro-2-nitrobenzoic acid
CAS 2516-95-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H4ClNO4
Linear Formula ClC6H3(NO2)CO2H
Formula Weight 201.57
MDL Number MFCD00007290
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents