Please login to your online account to display your discounted pricing

5-chloro-2-nitrobenzaldehyde, 97%, ACROS Organics™

$141.65 - $141.65

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-nitrobenzaldehyde
CAS 6628-86-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H4ClNO3
Linear Formula ClC6H3(NO2)CHO
Formula Weight 185.57
MDL Number MFCD00007289
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC329760250

  SDS
thermo fisher scientific
AC329760250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC329760050

  SDS
thermo fisher scientific
329760050
Glass bottle 5g Each for $141.65
Description & Specifications

Specifications

Physical Form Powder
Assay ≥96.0%
Formula Weight 185.57
Quantity 25g
Color Yellow
Melting Point 76° to 80°C
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Linear Formula ClC6H3(NO2)CHO
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 5-Chloro-2-nitrobenzaldehyde
Assay Percent Range 97%
Molecular Formula C7H4ClNO3
MDL Number MFCD00007289
CAS 6628-86-0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.