missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-2-nitroaniline, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 246950050

 View more versions of this product

Catalog No. AC246950050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

5-Chloro-2-nitroaniline
Glass bottle
126.0°C to 129.0°C
97%
96.0
96% min. (GC)
ClC6H3(NO2)NH2
12, 730
Solubility in water: 1.22g/L (20°C)
C1=CC(=C(C=C1Cl)N)[N+](=O)[O-]
172.57
172.57
97%
Authentic
Colorless to Yellow
5g
1635-61-6
100.0
C6H5ClN2O2
MFCD00007776
2-amino-4-chloro-nitrobenzene, 2-nitro-5-chloroaniline, 3-chloro-6-nitroaniline, 5-chloro-2-nitro aniline, 5-chloro-2-nitrobenzenamine, 5-chloro-2-nitrobenzeneamine, aniline, 5-chloro-2-nitro, benzenamine, 5-chloro-2-nitro, ccris 8934, unii-656xjm4con
ZCWXYZBQDNFULS-UHFFFAOYSA-N
5-chloro-2-nitroaniline
74218
Liquid

Chemical Identifiers

1635-61-6
172.57
ZCWXYZBQDNFULS-UHFFFAOYSA-N
74218
C1=CC(=C(C=C1Cl)N)[N+](=O)[O-]
C6H5ClN2O2
MFCD00007776
2-amino-4-chloro-nitrobenzene, 2-nitro-5-chloroaniline, 3-chloro-6-nitroaniline, 5-chloro-2-nitro aniline, 5-chloro-2-nitrobenzenamine, 5-chloro-2-nitrobenzeneamine, aniline, 5-chloro-2-nitro, benzenamine, 5-chloro-2-nitro, ccris 8934, unii-656xjm4con
5-chloro-2-nitroaniline
Safety and Handling

Safety and Handling

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 216-661-5

SDS Documents