Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-methylphenyl isocyanate, 98%

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-methylphenyl isocyanate
CAS 40411-27-6
Molecular Formula C8H6ClNO
Formula Weight 167.6
MDL Number MFCD00019913
UN Number UN2206
Beilstein 2691112
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1156703

 
alfa aesar™
L1156703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1156706

 
alfa aesar™
L1156706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 167.6
Chemical Name or Material 5-Chloro-2-methylphenyl isocyanate
Assay 98%
Molecular Formula C8H6ClNO
MDL Number MFCD00019913
CAS 40411-27-6
UN Number UN2206
Beilstein 2691112
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H330-H334-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P260-P304+P340-P305+P351+P338-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Flash Point 108°C (226°F)
Boiling Point 115°C (5mmHg)
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P304+P340-P305+P351+P338-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H330-H334-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C