missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-methylphenyl isocyanate, 98%

Manufacturer:  Alfa Aesar™ L1156706

 View more versions of this product

Catalog No. AAL1156706



Specifications

Specifications

5-Chloro-2-methylphenyl isocyanate
115°C (5mmHg)
40411-27-6
MFCD00019913
2691112
XEMUTFNBAICJEO-UHFFFAOYSA-N
4-chloro-2-isocyanato-1-methylbenzene
2769632
98%
108°C (226°F)
5g
C8H6ClNO
UN2206
5-chloro-2-methylphenyl isocyanate, 5-chloro-2-methylphenylisocyanate, benzene,4-chloro-2-isocyanato-1-methyl, benzene, 4-chloro-2-isocyanato-1-methyl, 5-chloro-2-methylbenzenisocyanate, acmc-1aqw4, 4-chloro-2-isocyanatotoluene, 2-methyl-5-chlorophenylisocyanate, 5-chloro-1-isocyanato-2-methyl benzene, 4-chloranyl-2-isocyanato-1-methyl-benzene
CC1=C(C=C(C=C1)Cl)N=C=O
167.592
167.6

Chemical Identifiers

40411-27-6
167.592
XEMUTFNBAICJEO-UHFFFAOYSA-N
2769632
CC1=C(C=C(C=C1)Cl)N=C=O
C8H6ClNO
MFCD00019913
5-chloro-2-methylphenyl isocyanate, 5-chloro-2-methylphenylisocyanate, benzene,4-chloro-2-isocyanato-1-methyl, benzene, 4-chloro-2-isocyanato-1-methyl, 5-chloro-2-methylbenzenisocyanate, acmc-1aqw4, 4-chloro-2-isocyanatotoluene, 2-methyl-5-chlorophenylisocyanate, 5-chloro-1-isocyanato-2-methyl benzene, 4-chloranyl-2-isocyanato-1-methyl-benzene
4-chloro-2-isocyanato-1-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H330-H334-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P304+P340-P305+P351+P338-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents