Please login to your online account to display your discounted pricing

5-chloro-2-methylindole, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-chloro-2-methylindole
CAS 1075-35-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C9H8ClN
Formula Weight 165.62
MDL Number MFCD00005619
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC109410010

  SDS
thermo fisher scientific
109410010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder or Crystals
Assay ≥94%
Formula Weight 165.62
Quantity 1g
Color Light pink to beige-pale brown
Melting Point 112° to 118°C
Beilstein 20, 314
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 5-chloro-2-methylindole
Assay Percent Range 95%
Molecular Formula C9H8ClN
CAS 1075-35-0
Infrared Spectrum Authentic
MDL Number MFCD00005619
Packaging Glass bottle