Please login to your online account to display your discounted pricing

5-chloro-2-methylindole, 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  109410010

 View more versions of this product

Catalog No. AC109410010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder or Crystals
Assay ≥94%
Formula Weight 165.62
Quantity 1g
Color Light pink to beige-pale brown
Melting Point 112° to 118°C
Beilstein 20, 314
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 5-chloro-2-methylindole
Assay Percent Range 95%
Molecular Formula C9H8ClN
CAS 1075-35-0
Infrared Spectrum Authentic
MDL Number MFCD00005619
Packaging Glass bottle

Specifications

Chemical Name or Material 5-chloro-2-methylindole
CAS 1075-35-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C9H8ClN
Formula Weight 165.62
MDL Number MFCD00005619