Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-methylbenzothiazole, 98%

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-methylbenzothiazole
CAS 1006-99-1
Formula Weight 183.66
MDL Number MFCD00022881
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5574303

  SDS
alfa aesar™
H5574303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5574306

  SDS
alfa aesar™
H5574306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5574314

  SDS
alfa aesar™
H5574314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 65° to 70°C
Formula Weight 183.66
RTECS Number DL3325000
Assay 98%
Physical Form Solid
Color White to pale brown
CAS 1006-99-1
Quantity 1g
Chemical Name or Material 5-Chloro-2-methylbenzothiazole
TSCA TSCA
MDL Number MFCD00022881