missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-methylbenzoic acid, 98%

$17.88 - $70.03

Chemical Identifiers

CAS 7499-06-1
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00045842
InChI Key CSAPESWNZDOAFU-UHFFFAOYSA-N
Synonym 2-methyl-5-chlorobenzoic acid, 5-chloro-o-toluic acid, 5-chloro-2-methyl-benzoic acid, benzoic acid, 5-chloro-2-methyl, o-toluic acid, 5-chloro, 5-chloro-2-methylbenzoicacid, 5-chloro-2-methyl benzoic acid, 3-chloro-6-methylbenzoic acid, pubchem4578, ksc173q5r
PubChem CID 82009
IUPAC Name 5-chloro-2-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)Cl)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6064903 View Documents Alfa Aesar™
H6064903
1g Each for $17.88
 
AAH6064906 View Documents Alfa Aesar™
H6064906
5g Each for $70.03
 
Specifications

Chemical Identifiers

7499-06-1
170.59
CSAPESWNZDOAFU-UHFFFAOYSA-N
82009
CC1=C(C=C(C=C1)Cl)C(=O)O
C8H7ClO2
MFCD00045842
2-methyl-5-chlorobenzoic acid, 5-chloro-o-toluic acid, 5-chloro-2-methyl-benzoic acid, benzoic acid, 5-chloro-2-methyl, o-toluic acid, 5-chloro, 5-chloro-2-methylbenzoicacid, 5-chloro-2-methyl benzoic acid, 3-chloro-6-methylbenzoic acid, pubchem4578, ksc173q5r
5-chloro-2-methylbenzoic acid

Specifications

168°C to 173°C
7499-06-1
MFCD00045842
CSAPESWNZDOAFU-UHFFFAOYSA-N
5-chloro-2-methylbenzoic acid
82009
98%
1g
C8H7ClO2
2-methyl-5-chlorobenzoic acid, 5-chloro-o-toluic acid, 5-chloro-2-methyl-benzoic acid, benzoic acid, 5-chloro-2-methyl, o-toluic acid, 5-chloro, 5-chloro-2-methylbenzoicacid, 5-chloro-2-methyl benzoic acid, 3-chloro-6-methylbenzoic acid, pubchem4578, ksc173q5r
CC1=C(C=C(C=C1)Cl)C(=O)O
170.59
170.59
5-Chloro-2-methylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning