Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-methylaniline, 99%

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-methylaniline
CAS 95-79-4
Molecular Formula C7H8ClN
Formula Weight 141.6
MDL Number MFCD00007779
UN Number UN2239
Beilstein 878504
Synonym 2-Amino-4-chlorotoluene; 5-Chloro-o-toluidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2139222

  SDS
alfa aesar™
B2139222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2139236

  SDS
alfa aesar™
B2139236
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 141.6
Molecular Formula C7H8ClN
Synonym 2-Amino-4-chlorotoluene; 5-Chloro-o-toluidine
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROTOLUIDINES, SOLID
Flash Point 160°C (320°F)
Chemical Name or Material 5-Chloro-2-methylaniline
TSCA TSCA
Assay 99%
MDL Number MFCD00007779
CAS 95-79-4
UN Number UN2239
Beilstein 878504
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H350-H302-H319
May cause cancer.
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P281-P305+P351+P338-P301+P312-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number XU5075000
Boiling Point 236° to 238°C
Melting Point 21° to 24°C
Quantity 100g
Safety and Handling

Danger

GHS P Statement
P280-P281-P305+P351+P338-P301+P312-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H302-H319
May cause cancer.
Harmful if swallowed.
Causes serious eye irritation.