Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-methylaminobenzophenone, 99%

$41.14 - $41.14

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-methylaminobenzophenone
CAS 1022-13-5
Molecular Formula C14H12ClNO
Formula Weight 245.71
MDL Number MFCD00008284
Beilstein 882187
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0633809

  SDS
alfa aesar™
L0633809
10g Each for $41.14
Description & Specifications

Specifications

Formula Weight 245.71
Beilstein 882187
Quantity 10g
MDL Number MFCD00008284
Molecular Formula C14H12ClNO
Chemical Name or Material 5-Chloro-2-methylaminobenzophenone
Assay 99%
CAS 1022-13-5
RTECS Number DJ0414000
Melting Point 93° to 98°C
TSCA TSCA