missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-2-methoxypyridine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC453730050

 View more versions of this product

Catalog No. AC453730050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

5-Chloro-2-methoxypyridine
53°C
180°C to 184°C
13473-01-3
C6H6ClNO
NPYYXUYLIHZYOU-UHFFFAOYSA-N
5-chloro-2-methoxypyridine
4738276
97%
1.1930g/mL
1.193
5g
96% min. (GC)
2-methoxy-5-chloro pyridine, 2-methoxy-5-chloropyridine, 5-chloranyl-2-methoxy-pyridine, 5-chloro-2-methoxy-pyridine, 5-chloro-2-methoxypyridine, acmc-1c02r, ksc494g7j, pubchem6602, pyridine, 5-chloro-2-methoxy
COC1=NC=C(C=C1)Cl
143.57
143.57

Chemical Identifiers

13473-01-3
143.57
2-methoxy-5-chloro pyridine, 2-methoxy-5-chloropyridine, 5-chloranyl-2-methoxy-pyridine, 5-chloro-2-methoxy-pyridine, 5-chloro-2-methoxypyridine, acmc-1c02r, ksc494g7j, pubchem6602, pyridine, 5-chloro-2-methoxy
5-chloro-2-methoxypyridine
C6H6ClNO
NPYYXUYLIHZYOU-UHFFFAOYSA-N
4738276
COC1=NC=C(C=C1)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Warning

SDS Documents