missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-chloro-2-methoxyphenylboronic acid, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC434480010

 View more versions of this product

Catalog No. AC434480010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

5-chloro-2-methoxyphenylboronic acid
Glass bottle
1g
96.0
97%
MFCD01318966
FMBVAOHFMSQDGT-UHFFFAOYSA-N
(5-chloro-2-methoxyphenyl)boronic acid
2735751
Crystalline Powder
Authentic
White
89694-48-4
100.0
C7H8BClO3
5-chloro-2-methoxyphenyl boronic acid, 5-chloro-2-methoxybenzeneboronic acid, 5-chloro-2-methoxyphenyl boranediol, 2-methoxy-5-chlorophenylboronic acid, 5-chloro-2-methoxyphenyl boronic acid, 4-chloroanisole-2-boronic acid, boronic acid, 5-chloro-2-methoxyphenyl, 5-chloro-2-methoxyphenylboronicacid, pubchem1782, 5-chloro-2-methoxy-phenyl boronic acid
B(C1=C(C=CC(=C1)Cl)OC)(O)O
186.398
186.4
≥96.0%

Chemical Identifiers

89694-48-4
186.398
FMBVAOHFMSQDGT-UHFFFAOYSA-N
2735751
B(C1=C(C=CC(=C1)Cl)OC)(O)O
C7H8BClO3
MFCD01318966
5-chloro-2-methoxyphenyl boronic acid, 5-chloro-2-methoxybenzeneboronic acid, 5-chloro-2-methoxyphenyl boranediol, 2-methoxy-5-chlorophenylboronic acid, 5-chloro-2-methoxyphenyl boronic acid, 4-chloroanisole-2-boronic acid, boronic acid, 5-chloro-2-methoxyphenyl, 5-chloro-2-methoxyphenylboronicacid, pubchem1782, 5-chloro-2-methoxy-phenyl boronic acid
(5-chloro-2-methoxyphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents