missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-chloro-2-methoxyphenylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 89694-48-4
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H8BClO3
Formula Weight 186.4
MDL Number MFCD01318966
Chemical Name or Material 5-chloro-2-methoxyphenylboronic acid
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC434480010 SDS Thermo Fisher Scientific
AC434480010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

89694-48-4
100.0
C7H8BClO3
MFCD01318966
≥96.0%
96.0
97%
186.4
5-chloro-2-methoxyphenylboronic acid

Specifications

186.4
1g
96.0
89694-48-4
97%
Glass bottle
5-chloro-2-methoxyphenylboronic acid
Crystalline Powder
White
100.0
C7H8BClO3
MFCD01318966
Authentic
≥96.0%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.