Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-methoxyphenyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-methoxyphenyl isocyanate
CAS 55440-54-5
Molecular Formula C8H6ClNO2
Formula Weight 183.6
MDL Number MFCD00013860
UN Number UN2811
Beilstein 2719682
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2239509

  SDS
alfa aesar™
B2239509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 183.6
Quantity 10g
Chemical Name or Material 5-Chloro-2-methoxyphenyl isocyanate
Melting Point 58° to 62°C
Beilstein 2719682
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Molecular Formula C8H6ClNO2
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN2811
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
MDL Number MFCD00013860
CAS 55440-54-5
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.