Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-methoxybenzonitrile, 98%

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-methoxybenzonitrile
CAS 55877-79-7
Molecular Formula C8H6ClNO
Formula Weight 167.6
MDL Number MFCD00052867
UN Number UN3439
Beilstein 3245801
Synonym 5-Chloro-o-anisonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1518503

  SDS
alfa aesar™
A1518503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1518506

  SDS
alfa aesar™
A1518506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 167.6
Chemical Name or Material 5-Chloro-2-methoxybenzonitrile
Assay 98%
Molecular Formula C8H6ClNO
MDL Number MFCD00052867
CAS 55877-79-7
UN Number UN3439
Beilstein 3245801
Synonym 5-Chloro-o-anisonitrile
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Melting Point 98° to 102°C
Quantity 1g

Applications
It is employed as an intermediate for pharmaceutical.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.