missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-methoxybenzonitrile, 98%

Manufacturer:  Alfa Aesar™ A1518503

 View more versions of this product

Catalog No. AAA1518503



Description

Description

Applications
It is employed as an intermediate for pharmaceutical.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

5-Chloro-2-methoxybenzonitrile
1g
C8H6ClNO
UN3439
2-methoxy-5-chlorobenzonitrile, 4-chloro-2-cyanoanisole, 5-chloro-2-methoxy-benzonitrile, benzonitrile, 5-chloro-2-methoxy, 5-chloro-2-methoxybenzenecarbonitrile, pubchem3662, acmc-1avm0, ksc490q4j, 5-chloro-o-anisonitrile, lreabokklivxna-uhfffaoysa
COC1=C(C=C(C=C1)Cl)C#N
167.592
167.6
98°C to 102°C
55877-79-7
MFCD00052867
3245801
LREABOKKLIVXNA-UHFFFAOYSA-N
5-chloro-2-methoxybenzonitrile
2800979
98%

Chemical Identifiers

55877-79-7
167.592
LREABOKKLIVXNA-UHFFFAOYSA-N
2800979
COC1=C(C=C(C=C1)Cl)C#N
C8H6ClNO
MFCD00052867
2-methoxy-5-chlorobenzonitrile, 4-chloro-2-cyanoanisole, 5-chloro-2-methoxy-benzonitrile, benzonitrile, 5-chloro-2-methoxy, 5-chloro-2-methoxybenzenecarbonitrile, pubchem3662, acmc-1avm0, ksc490q4j, 5-chloro-o-anisonitrile, lreabokklivxna-uhfffaoysa
5-chloro-2-methoxybenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents