Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-methoxybenzoic acid, 99%

$78.20 - $842.28

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-methoxybenzoic acid
CAS 3438-16-2
Molecular Formula C8H7ClO3
Formula Weight 186.59
MDL Number MFCD00017546
Synonym 5-Chloro-o-anisic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2497414

  SDS
alfa aesar™
B2497414
25g Each for $78.20

AAB2497422

  SDS
alfa aesar™
B2497422
100g Each for $198.89

AAB2497436

  SDS
alfa aesar™
B2497436
500g Each for $842.28
Description & Specifications

Specifications

Formula Weight 186.59
Synonym 5-Chloro-o-anisic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 97° to 100°C
Assay 99%
CAS 3438-16-2
Quantity 25g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H7ClO3
Chemical Name or Material 5-Chloro-2-methoxybenzoic acid
MDL Number MFCD00017546
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.