Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-methoxyaniline hydrochloride, 98%

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-methoxyaniline hydrochloride
CAS 4274-03-7
Molecular Formula C7H8ClNO·HCl
Formula Weight 194.06
MDL Number MFCD00035488
UN Number UN2811
Synonym 2-Amino-4-chloroanisole hydrochloride; 5-Chloro-o-anisidine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1288722

  SDS
alfa aesar™
A1288722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1288736

  SDS
alfa aesar™
A1288736
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.06
UN Number UN2811
TSCA TSCA
Melting Point ≈228°C (dec.)
MDL Number MFCD00035488
Chemical Name or Material 5-Chloro-2-methoxyaniline hydrochloride
Molecular Formula C7H8ClNO·HCl
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Synonym 2-Amino-4-chloroanisole hydrochloride; 5-Chloro-o-anisidine hydrochloride
Health Hazard 3 GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Health Hazard 2 GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
Quantity 100g
CAS 4274-03-7
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.