Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-methoxy-4-methylbenzenesulfonamide, 98%

$76.39 - $262.18

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-methoxy-4-methylbenzenesulfonamide
CAS 199590-75-5
Molecular Formula C8H10ClNO3S
Formula Weight 253.69
MDL Number MFCD08445708
Synonym 5-Chloro-2-methoxy-p-toluenesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2601903

  SDS
alfa aesar™
H2601903
1g Each for $76.39

AAH2601906

  SDS
alfa aesar™
H2601906
5g Each for $262.18
Description & Specifications

Specifications

Formula Weight 253.69
Assay 98%
Molecular Formula C8H10ClNO3S
MDL Number MFCD08445708
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Chemical Name or Material 5-Chloro-2-methoxy-4-methylbenzenesulfonamide
CAS 199590-75-5
Synonym 5-Chloro-2-methoxy-p-toluenesulfonamide
Health Hazard 1 Warning
Melting Point 169° to 171°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.