Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-iodotoluene, 97%

$55.55 - $55.55

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-iodotoluene
CAS 23399-70-4
Molecular Formula C7H6ClI
Formula Weight 252.48
MDL Number MFCD00060665
Synonym 4-Chloro-1-iodo-2-methylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2168103

 
alfa aesar™
B2168103
1g Each for $55.55
Description & Specifications

Specifications

Formula Weight 252.48
Chemical Name or Material 5-Chloro-2-iodotoluene
Assay 97%
Molecular Formula C7H6ClI
MDL Number MFCD00060665
CAS 23399-70-4
Synonym 4-Chloro-1-iodo-2-methylbenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 120° to 122°C (15mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.